Anti-IL-15 Rabbit Polyclonal Antibody

Supplier: Peprotech
500-P173-50UG 500-P173-100UG 500-P173-1MG
10776-598EA 595.93 CAD
10776-598 10776-600 10776-602
Anti-IL-15 Rabbit Polyclonal Antibody
Antibodies
Produced from sera of rabbits immunized with highly pure Recombinant Murine IL-15. Anti­Murine IL-15­specific antibody was purified by affinity chromatography employing an immobilized Murine IL-15 matrix.

Applications tested can vary by lot number. See Product Data Sheet for more information.

Type: Primary
Antigen: IL-15
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Murine
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR