Anti-IL-10 Rabbit Polyclonal Antibody

Supplier: Peprotech
500-P60-50UG 500-P60-100UG 500-P60-1MG
10775-502EA 560.17 CAD
10775-502 10775-504 10775-506
Anti-IL-10 Rabbit Polyclonal Antibody
Antibodies
Produced from sera of rabbits immunized with highly pure Recombinant Murine IL-10. Anti­Murine IL-10­specific antibody was purified by affinity chromatography employing an immobilized Murine IL-10 matrix.

Applications tested can vary by lot number. See Product Data Sheet for more information.

Type: Primary
Antigen: IL-10
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Murine
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR