Anti-Exodus-2 (CCL21) Rabbit Polyclonal Antibody

Supplier: Peprotech
500-P114-50UG 500-P114-100UG 500-P114-1MG
10776-058EA 581.63 CAD
10776-058 10776-060 10776-062
Anti-Exodus-2 (CCL21) Rabbit Polyclonal Antibody
Antibodies
Produced from sera of rabbits immunized with highly pure Recombinant Murine Exodus-2 (CCL21). Anti­Murine Exodus-2 (CCL21)­specific antibody was purified by affinity chromatography employing an immobilized Murine Exodus-2 matrix.

Applications tested can vary by lot number. See Product Data Sheet for more information.

Type: Primary
Antigen: Exodus-2 (CCL21)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Murine
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR