Anti-TNF-beta Rabbit Polyclonal Antibody

Supplier: Peprotech
500-P31B-50UG 500-P31B-100UG 500-P31B-1MG
10775-194EA 560.17 CAD
10775-194 10775-196 10775-198
Anti-TNF-beta Rabbit Polyclonal Antibody
Antibodies
Produced from sera of rabbits immunized with highly pure Recombinant Human TNF-β. Anti­Human TNF-β­specific antibody was purified by affinity chromatography employing an immobilized Human TNF-β matrix.

Applications tested can vary by lot number. See Product Data Sheet for more information.

Type: Primary
Antigen: TNF-beta
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR