Anti-TECK (CCL25) Rabbit Polyclonal Antibody

Supplier: Peprotech
500-P134-50UG 500-P134-100UG 500-P134-1MG
10771-376EA 560.17 CAD
10771-376 10771-378 10771-380
Anti-TECK (CCL25) Rabbit Polyclonal Antibody
Antibodies
Produced from sera of rabbits immunized with highly pure Recombinant Human TECK (CCL25). Anti­Human TECK (CCL25)­specific antibody was purified by affinity chromatography employing an immobilized Human TECK matrix.

Applications tested can vary by lot number. See Product Data Sheet for more information.

Type: Primary
Antigen: TECK (CCL25)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR