Anti-PTHrP Rabbit Polyclonal Antibody

Supplier: Peprotech
500-P276-50UG 500-P276-100UG 500-P276-1MG
10777-466EA 536.88 CAD
10777-466 10777-468 10777-470
Anti-PTHrP Rabbit Polyclonal Antibody
Antibodies
Produced from sera of rabbits immunized with highly pure Recombinant Human PTHrP. Anti­Human PTHrP­specific antibody was purified by affinity chromatography employing an immobilized Human PTHrP matrix.

Applications tested can vary by lot number. See Product Data Sheet for more information.

Type: Primary
Antigen: PTHrP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR