Anti-PlGF-1 Rabbit Polyclonal Antibody

Supplier: Peprotech
500-P226-50UG 500-P226-100UG 500-P226-1MG
10777-026EA 536.88 CAD
10777-026 10777-028 10777-030
Anti-PlGF-1 Rabbit Polyclonal Antibody
Antibodies
Produced from sera of rabbits immunized with highly pure Recombinant Human PlGF-1. Anti­Human PlGF-1­specific antibody was purified by affinity chromatography employing an immobilized Human PlGF-1 matrix.

Applications tested can vary by lot number. See Product Data Sheet for more information.

Type: Primary
Antigen: PlGF-1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR