Anti-NAP-2 (CXCL7) Rabbit Polyclonal Antibody

Supplier: Peprotech
500-P03-50UG 500-P03-100UG 500-P03-1MG
10774-946EA 536.88 CAD
10774-946 10774-948 10774-950
Anti-NAP-2 (CXCL7) Rabbit Polyclonal Antibody
Antibodies
Produced from sera of rabbits immunized with highly pure Recombinant Human NAP-2 (CXCL7). Anti­Human NAP-2 (CXCL7)­specific antibody was purified by affinity chromatography employing an immobilized Human NAP-2 matrix.

Applications tested can vary by lot number. See Product Data Sheet for more information.

Type: Primary
Antigen: NAP-2 (CXCL7)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR