Anti-MIP-4 (CCL18) Rabbit Polyclonal Antibody

Supplier: Peprotech
500-P108-50UG 500-P108-100UG 500-P108-1MG
10771-360EA 581.63 CAD
10771-360 10771-362 10771-364
Anti-MIP-4 (CCL18) Rabbit Polyclonal Antibody
Antibodies
Produced from sera of rabbits immunized with highly pure Recombinant Human MIP-4 (CCL18). Anti­Human MIP-4 (CCL18)­specific antibody was purified by affinity chromatography employing an immobilized Human MIP-4 matrix.

Applications tested can vary by lot number. See Product Data Sheet for more information.

Type: Primary
Antigen: MIP-4 (CCL18)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR