Anti-MIP-3alpha (CCL20) Rabbit Polyclonal Antibody

Supplier: Peprotech
500-P95A-50UG 500-P95A-1MG
10775-906EA 536.88 CAD
10775-906 10775-910
Anti-MIP-3alpha (CCL20) Rabbit Polyclonal Antibody
Antibodies
Produced from sera of rabbits immunized with highly pure Recombinant Human MIP-3α (CCL20). Anti­Human MIP-3α (CCL20)­specific antibody was purified by affinity chromatography employing an immobilized Human MIP-3α matrix.

Applications tested can vary by lot number. See Product Data Sheet for more information.

Type: Primary
Antigen: MIP-3alpha (CCL20)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR