Anti-Klotho Rabbit Polyclonal Antibody

Supplier: Peprotech
500-P296-50UG 500-P296-100UG 500-P296-1MG
10777-546EA 581.63 CAD
10777-546 10777-548 10777-550
Anti-Klotho Rabbit Polyclonal Antibody
Antibodies
Produced from sera of rabbits immunized with highly pure Recombinant Human Klotho. Anti­Human Klotho­specific antibody was purified by affinity chromatography employing an immobilized Human Klotho matrix.

Applications tested can vary by lot number. See Product Data Sheet for more information.

Type: Primary
Antigen: Klotho
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR