Anti-IL-8 (CXCL8) Rabbit Polyclonal Antibody

Supplier: Peprotech
500-P28-50UG 500-P28-100UG 500-P28-1MG
10775-134EA 560.17 CAD
10775-134 10775-136 10775-138
Anti-IL-8 (CXCL8) Rabbit Polyclonal Antibody
Antibodies
Produced from sera of rabbits immunized with highly pure Recombinant Human IL-8 (CXCL8). Anti­Human IL-8 (CXCL8)­specific antibody was purified by affinity chromatography employing an immobilized Human IL-8 matrix.

Applications tested can vary by lot number. See Product Data Sheet for more information.

Type: Primary
Antigen: IL-8 (CXCL8)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR