Anti-IL-4 Rabbit Polyclonal Antibody

Supplier: Peprotech
500-P24-50UG 500-P24-100UG 500-P24-1MG
10771-224EA 560.17 CAD
10771-224 10771-226 10771-228
Anti-IL-4 Rabbit Polyclonal Antibody
Antibodies
Produced from sera of rabbits immunized with highly pure Recombinant Human IL-4. Anti­Human IL-4­specific antibody was purified by affinity chromatography employing an immobilized Human IL-4 matrix.

Applications tested can vary by lot number. See Product Data Sheet for more information.

Type: Primary
Antigen: IL-4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR