Anti-IL-33 Rabbit Polyclonal Antibody

Supplier: Peprotech
500-P261-50UG 500-P261-100UG 500-P261-1MG
10777-370EA 571.15 CAD
10777-370 10777-372 10777-374
Anti-IL-33 Rabbit Polyclonal Antibody
Antibodies
Produced from sera of rabbits immunized with highly pure Recombinant Human IL-33. Anti­Human IL-33­specific antibody was purified by affinity chromatography employing an immobilized Human IL-33 matrix.

Applications tested can vary by lot number. See Product Data Sheet for more information.

Type: Primary
Antigen: IL-33
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR