Anti-IL-22 Rabbit Polyclonal Antibody

Supplier: Peprotech
500-P211-50UG 500-P211-100UG 500-P211-1MG
10771-488EA 560.17 CAD
10771-488 10771-490 10771-492
Anti-IL-22 Rabbit Polyclonal Antibody
Antibodies
Produced from sera of rabbits immunized with highly pure Recombinant Human IL-22. Anti­Human IL-22­specific antibody was purified by affinity chromatography employing an immobilized Human IL-22 matrix.

Applications tested can vary by lot number. See Product Data Sheet for more information.

Type: Primary
Antigen: IL-22
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR