Anti-IL-17F Rabbit Polyclonal Antibody

Supplier: Peprotech
500-P90-50UG 500-P90-100UG 500-P90-1MG
10775-846EA 560.17 CAD
10775-846 10775-848 10775-850
Anti-IL-17F Rabbit Polyclonal Antibody
Antibodies
Produced from sera of rabbits immunized with highly pure Recombinant Human IL-17F. Anti­Human IL-17F­specific antibody was purified by affinity chromatography employing an immobilized Human IL-17F matrix.

Applications tested can vary by lot number. See Product Data Sheet for more information.

Type: Primary
Antigen: IL-17F
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR