Anti-IL-17B Rabbit Polyclonal Antibody

Supplier: Peprotech
500-P248-50UG 500-P248-100UG 500-P248-1MG
10777-230EA 536.88 CAD
10777-230 10777-232 10777-234
Anti-IL-17B Rabbit Polyclonal Antibody
Antibodies
Produced from sera of rabbits immunized with highly pure Recombinant Human IL-17B. Anti­Human IL-17B­specific antibody was purified by affinity chromatography employing an immobilized Human IL-17B matrix.

Applications tested can vary by lot number. See Product Data Sheet for more information.

Type: Primary
Antigen: IL-17B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR