Anti-IL-13 Rabbit Polyclonal Antibody

Supplier: Peprotech
500-P13-50UG 500-P13-100UG 500-P13-1MG
10775-034EA 560.17 CAD
10775-034 10775-036 10775-038
Anti-IL-13 Rabbit Polyclonal Antibody
Antibodies
Produced from sera of rabbits immunized with highly pure Recombinant Human IL-13. Anti­Human IL-13­specific antibody was purified by affinity chromatography employing an immobilized Human IL-13 matrix.

Applications tested can vary by lot number. See Product Data Sheet for more information.

Type: Primary
Antigen: IL-13
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR