Anti-IL-1RA Rabbit Polyclonal Antibody

Supplier: Peprotech
500-P209-100UG 500-P209-1MG
10776-892EA 686.72 CAD
10776-892 10776-894
Anti-IL-1RA Rabbit Polyclonal Antibody
Antibodies
Produced from sera of rabbits immunized with highly pure Recombinant Human IL-1RA. Anti­Human IL-1RA­specific antibody was purified by affinity chromatography employing an immobilized Human IL-1RA matrix.

Applications tested can vary by lot number. See Product Data Sheet for more information.

Type: Primary
Antigen: IL-1RA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR