Anti-IGF-BP3 Rabbit Polyclonal Antibody

Supplier: Peprotech
500-P230-50UG 500-P230-100UG 500-P230-1MG
10777-062EA 536.88 CAD
10777-062 10777-064 10777-066
Anti-IGF-BP3 Rabbit Polyclonal Antibody
Antibodies
Produced from sera of rabbits immunized with highly pure Recombinant Human IGF-BP3. Anti­Human IGF-BP3­specific antibody was purified by affinity chromatography employing an immobilized Human IGF-BP3 matrix.

Applications tested can vary by lot number. See Product Data Sheet for more information.

Type: Primary
Antigen: IGF-BP3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR