Anti-IFN-lambda2 Rabbit Polyclonal Antibody

Supplier: Peprotech
500-P247-50UG 500-P247-100UG 500-P247-1MG
10777-218EA 536.88 CAD
10777-218 10777-220 10777-222
Anti-IFN-lambda2 Rabbit Polyclonal Antibody
Antibodies
Produced from sera of rabbits immunized with highly pure Recombinant Human IFN-λ2. Anti­Human IFN-λ2­specific antibody was purified by affinity chromatography employing an immobilized Human IFN-λ2 matrix.

Applications tested can vary by lot number. See Product Data Sheet for more information.

Type: Primary
Antigen: IFN-lambda2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Human
Order Now

SPECIFICATIONS

Antigen symbol IFN-lambda2
Environmentally Preferable
Antigen name IFN-lambda2
Antibody type Primary
Clonality Polyclonal
Conjugation Unconjugated
Reactivity Human
Host Rabbit
Isotype IgG
Western blot Yes
ELISA Yes
Immunogen Recombinant Human IFN-λ2
Form Lyophilized
Molecular weight 19.6 kDa
Purification Recombinant Human IFN-λ2
Storage buffer See Product Data Sheet for more information.
Storage temperature Store at –20 °C prior to reconstitution.
Shipping temperature Ambient

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR