Anti-GRO-beta (CXCL2) Rabbit Polyclonal Antibody

Supplier: Peprotech
500-P104-50UG 500-P104-100UG 500-P104-1MG
10775-986EA 581.63 CAD
10775-986 10775-988 10775-990
Anti-GRO-beta (CXCL2) Rabbit Polyclonal Antibody
Antibodies
Produced from sera of rabbits immunized with highly pure Recombinant Human GRO-β (CXCL2). Anti­Human GRO-β (CXCL2)­specific antibody was purified by affinity chromatography employing an immobilized Human GRO-β matrix.

Applications tested can vary by lot number. See Product Data Sheet for more information.

Type: Primary
Antigen: GRO-beta (CXCL2)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR