Anti-GDF-3 Rabbit Polyclonal Antibody

Supplier: Peprotech
500-P235-50UG 500-P235-100UG 500-P235-1MG
10777-098EA 536.88 CAD
10777-098 10777-100 10777-102
Anti-GDF-3 Rabbit Polyclonal Antibody
Antibodies
Produced from sera of rabbits immunized with highly pure Recombinant Human GDF-3. Anti­Human GDF-3­specific antibody was purified by affinity chromatography employing an immobilized Human GDF-3 matrix.

Applications tested can vary by lot number. See Product Data Sheet for more information.

Type: Primary
Antigen: GDF-3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR