Anti-Follistatin Rabbit Polyclonal Antibody

Supplier: Peprotech
500-P207-50UG 500-P207-100UG 500-P207-1MG
10776-878EA 536.88 CAD
10776-878 10776-880 10776-882
Anti-Follistatin Rabbit Polyclonal Antibody
Antibodies
Produced from sera of rabbits immunized with highly pure Recombinant Human Follistatin. Anti­Human Follistatin­specific antibody was purified by affinity chromatography employing an immobilized Human Follistatin matrix.

Applications tested can vary by lot number. See Product Data Sheet for more information.

Type: Primary
Antigen: Follistatin
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR