Anti-Flt3-Ligand Rabbit Polyclonal Antibody

Supplier: Peprotech
500-P42-50UG 500-P42-100UG 500-P42-1MG
10775-302EA 536.88 CAD
10775-302 10775-304 10775-306
Anti-Flt3-Ligand Rabbit Polyclonal Antibody
Antibodies
Produced from sera of rabbits immunized with highly pure Recombinant Human Flt3-Ligand. Anti­Human Flt3-Ligand­specific antibody was purified by affinity chromatography employing an immobilized Human Flt3-Ligand matrix.

Applications tested can vary by lot number. See Product Data Sheet for more information.

Type: Primary
Antigen: Flt3-Ligand
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR