Anti-FGF-17 Rabbit Polyclonal Antibody

Supplier: Peprotech
500-P152-50UG 500-P152-100UG 500-P152-1MG
10781-572EA 536.88 CAD
10781-572 10781-574 10781-576
Anti-FGF-17 Rabbit Polyclonal Antibody
Antibodies
Produced from sera of rabbits immunized with highly pure Recombinant Human FGF-17. Anti­Human FGF-17­specific antibody was purified by affinity chromatography employing an immobilized Human FGF-17 matrix.

Applications tested can vary by lot number. See Product Data Sheet for more information.

Type: Primary
Antigen: FGF-17
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR