Anti-sFas Receptor Rabbit Polyclonal Antibody

Supplier: Peprotech
500-P295-50UG 500-P295-100UG 500-P295-1MG
10777-534EA 581.63 CAD
10777-534 10777-536 10777-538
Anti-sFas Receptor Rabbit Polyclonal Antibody
Antibodies
Produced from sera of rabbits immunized with highly pure Recombinant Human sFas Receptor. Anti­Human sFas Receptor­specific antibody was purified by affinity chromatography employing an immobilized Human sFas Receptor matrix.

Applications tested can vary by lot number. See Product Data Sheet for more information.

Type: Primary
Antigen: sFas Receptor
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR