Anti-Exodus-2 (CCL21) Rabbit Polyclonal Antibody

Supplier: Peprotech
500-P109-50UG 500-P109-100UG 500-P109-1MG
10771-372EA 581.63 CAD
10771-372 10771-374 10776-002
Anti-Exodus-2 (CCL21) Rabbit Polyclonal Antibody
Antibodies
Produced from sera of rabbits immunized with highly pure Recombinant Human Exodus-2 (CCL21). Anti­Human Exodus-2 (CCL21)­specific antibody was purified by affinity chromatography employing an immobilized Human Exodus-2 matrix.

Applications tested can vary by lot number. See Product Data Sheet for more information.

Type: Primary
Antigen: Exodus-2 (CCL21)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Human
Order Now

SPECIFICATIONS

Antigen symbol Exodus-2 (CCL21)
Environmentally Preferable
Antigen name Exodus-2
Antibody type Primary
Clonality Polyclonal
Conjugation Unconjugated
Reactivity Human
Host Rabbit
Isotype IgG
Western blot Yes
ELISA Yes
Immunogen Recombinant Human Exodus-2 (CCL21)
Form Lyophilized
Molecular weight 12.2 kDa
Purification Recombinant Human Exodus-2 (CCL21)
Storage buffer See Product Data Sheet for more information.
Storage temperature Store at –20 °C prior to reconstitution.
Shipping temperature Ambient

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR