Anti-Eotaxin (CCL11) Rabbit Polyclonal Antibody

Supplier: Peprotech
500-P41-50UG 500-P41-100UG 500-P41-1MG
10775-278EA 536.88 CAD
10775-278 10775-280 10775-282
Anti-Eotaxin (CCL11) Rabbit Polyclonal Antibody
Antibodies
Produced from sera of rabbits immunized with highly pure Recombinant Human Eotaxin (CCL11). Anti­Human Eotaxin (CCL11)­specific antibody was purified by affinity chromatography employing an immobilized Human Eotaxin matrix.

Applications tested can vary by lot number. See Product Data Sheet for more information.

Type: Primary
Antigen: Eotaxin (CCL11)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR