Anti-Endostatin Rabbit Polyclonal Antibody

Supplier: PeproTech
500P2621MG 500P26250UG 500P262100UG
10777-386EA 4168.59 CAD
10777-386 10777-384 10777-382
Anti-Endostatin Rabbit Polyclonal Antibody
Antibodies
Produced from sera of rabbits immunized with highly pure Recombinant Human Endostatin. Anti­Human Endostatin­specific antibody was purified by affinity chromatography employing an immobilized Human Endostatin matrix.

Applications tested can vary by lot number. See Product Data Sheet for more information.

Type: Primary
Antigen: Endostatin
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR