Anti-EGF Receptor (EGFR) Rabbit Polyclonal Antibody

Supplier: Peprotech
500-P306-50UG 500-P306-100UG 500-P306-1MG
10781-628EA 536.88 CAD
10781-628 10781-630 10781-632
Anti-EGF Receptor (EGFR) Rabbit Polyclonal Antibody
Antibodies
Produced from sera of rabbits immunized with highly pure Recombinant Human EGF Receptor (EGFR). Anti­Human EGF Receptor (EGFR)­specific antibody was purified by affinity chromatography employing an immobilized Human EGF Receptor (EGFR) matrix.

Applications tested can vary by lot number. See Product Data Sheet for more information.

Type: Primary
Antigen: EGF Receptor (EGFR)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR