Anti-CTGFL/WISP-2 Rabbit Polyclonal Antibody

Supplier: Peprotech
500-P212-50UG 500-P212-100UG 500-P212-1MG
10776-918EA 536.88 CAD
10776-918 10776-920 10776-922
Anti-CTGFL/WISP-2 Rabbit Polyclonal Antibody
Antibodies
Produced from sera of rabbits immunized with highly pure Recombinant Human CTGFL/WISP-2. Anti­Human CTGFL/WISP-2­specific antibody was purified by affinity chromatography employing an immobilized Human CTGFL/WISP-2 matrix.

Applications tested can vary by lot number. See Product Data Sheet for more information.

Type: Primary
Antigen: CTGFL/WISP-2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR