Anti-beta-NGF Rabbit Polyclonal Antibody

Supplier: Peprotech
500-P85-50UG 500-P85-100UG 500-P85-1MG
10775-774EA 536.88 CAD
10775-774 10775-776 10775-778
Anti-beta-NGF Rabbit Polyclonal Antibody
Antibodies
Produced from sera of rabbits immunized with highly pure Recombinant Human β-NGF. Anti­Human β-NGF­specific antibody was purified by affinity chromatography employing an immobilized Human β-NGF matrix.

Applications tested can vary by lot number. See Product Data Sheet for more information.

Type: Primary
Antigen: β-NGF
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR