Anti-BD-4 Rabbit Polyclonal Antibody

Supplier: Peprotech
500-P268-50UG 500-P268-100UG 500-P268-1MG
10777-406EA 536.88 CAD
10777-406 10777-408 10777-410
Anti-BD-4 Rabbit Polyclonal Antibody
Antibodies
Produced from sera of rabbits immunized with highly pure Recombinant Human BD-4. Anti­Human BD-4­specific antibody was purified by affinity chromatography employing an immobilized Human BD-4 matrix.

Applications tested can vary by lot number. See Product Data Sheet for more information.

Type: Primary
Antigen: BD-4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR