Anti-sTRAIL Receptor-2 Rabbit Polyclonal Antibody

Supplier: Peprotech
500-P299-50UG 500-P299-100UG 500-P299-1MG
10777-570EA 536.88 CAD
10777-570 10777-572 10777-574
Anti-sTRAIL Receptor-2 Rabbit Polyclonal Antibody
Antibodies
Produced from sera of rabbits immunized with highly pure Recombinant Human sTRAIL Receptor-2. Anti­Human sTRAIL Receptor-2­specific antibody was purified by affinity chromatography employing an immobilized Human sTRAIL Receptor-2 matrix.

Applications tested can vary by lot number. See Product Data Sheet for more information.

Type: Primary
Antigen: sTRAIL Receptor-2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR