Anti-Osteopontin Rabbit Polyclonal Antibody

Supplier: Peprotech
500-P314-100UG 500-P314-1MG
75999-640EA 686.72 CAD
75999-640 75999-642
Anti-Osteopontin Rabbit Polyclonal Antibody
Antibodies
Produced from sera of rabbits immunized with highly pure Recombinant Human Osteopontin. Anti­Human Osteopontin­specific antibody was purified by affinity chromatography employing an immobilized Human Osteopontin matrix.

Applications tested can vary by lot number. See Product Data Sheet for more information.

Type: Primary
Antigen: Osteopontin
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR