Anti-TNF-alpha Goat Polyclonal Antibody (Biotin)

Supplier: Peprotech
500-P64GBT-25UG 500-P64GBT-50UG
10775-552EA 536.88 CAD
10775-552 10775-554 10775-556
Anti-TNF-alpha Goat Polyclonal Antibody (Biotin)
Antibodies
Produced from sera of goats immunized with highly pure Recombinant Murine TNF-α. Anti­Murine TNF-α­specific antibody was purified by affinity chromatography and then biotinylated.

Applications tested can vary by lot number. See Product Data Sheet for more information.

Type: Secondary
Antigen: TNF-alpha
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Goat
Isotype: Goat IgG
Reactivity: Murine
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR