Anti-EGF Goat Polyclonal Antibody (Biotin)

Supplier: Peprotech
500-P174GBT-25UG 500-P174GBT-50UG
10776-616EA 536.88 CAD
10776-616 10776-618 10776-620
Anti-EGF Goat Polyclonal Antibody (Biotin)
Antibodies
Produced from sera of goats immunized with highly pure Recombinant Murine EGF. Anti­Murine EGF­specific antibody was purified by affinity chromatography and then biotinylated.

Applications tested can vary by lot number. See Product Data Sheet for more information.

Type: Secondary
Antigen: EGF
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Goat
Isotype: Goat IgG
Reactivity: Murine
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR