Anti-LD78beta (CCL3L1) Goat Polyclonal Antibody (Biotin)

Supplier: PeproTech
500P187GBT1MG 500P187GBT25UG 500P187GBT50UG
10776-732EA 8330.9 CAD
10776-732 10776-728 10776-730
Anti-LD78beta (CCL3L1) Goat Polyclonal Antibody (Biotin)
Antibodies
Produced from sera of goats immunized with highly pure Recombinant Human LD78β (CCL3L1). Anti­Human LD78β (CCL3L1)­specific antibody was purified by affinity chromatography and then biotinylated.

Applications tested can vary by lot number. See Product Data Sheet for more information.

Type: Secondary
Antigen: LD78beta (CCL3L1)
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Goat
Isotype: Goat IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR