Anti-FGF-10 Goat Polyclonal Antibody (Biotin)

Supplier: Peprotech
500-P151GBT-25UG 500-P151GBT-50UG
10771-446EA 536.88 CAD
10771-446 10781-568 10781-570
Anti-FGF-10 Goat Polyclonal Antibody (Biotin)
Antibodies
Produced from sera of goats immunized with highly pure Recombinant Human FGF-10. Anti­Human FGF-10­specific antibody was purified by affinity chromatography and then biotinylated.

Applications tested can vary by lot number. See Product Data Sheet for more information.

Type: Secondary
Antigen: FGF-10
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Goat
Isotype: Goat IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR