Anti-Eotaxin (CCL11) Goat Polyclonal Antibody (Biotin)

Supplier: Peprotech
500-P41GBT-25UG 500-P41GBT-50UG
10775-296EA 536.88 CAD
10775-296 10775-298 10775-300
Anti-Eotaxin (CCL11) Goat Polyclonal Antibody (Biotin)
Antibodies
Produced from sera of goats immunized with highly pure Recombinant Human Eotaxin (CCL11). Anti­Human Eotaxin (CCL11)­specific antibody was purified by affinity chromatography and then biotinylated.

Applications tested can vary by lot number. See Product Data Sheet for more information.

Type: Secondary
Antigen: Eotaxin (CCL11)
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Goat
Isotype: Goat IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR