Anti-BD-2 Goat Polyclonal Antibody (Biotin)

Supplier: Peprotech
500-P161GBT-25UG 500-P161GBT-50UG
10776-504EA 560.17 CAD
10776-504 10776-506 10776-508
Anti-BD-2 Goat Polyclonal Antibody (Biotin)
Antibodies
Produced from sera of goats immunized with highly pure Recombinant Human BD-2. Anti­Human BD-2­specific antibody was purified by affinity chromatography and then biotinylated.

Applications tested can vary by lot number. See Product Data Sheet for more information.

Type: Secondary
Antigen: BD-2
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Goat
Isotype: Goat IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR