Anti-IFN-gamma Goat Polyclonal Antibody

Supplier: Peprotech
500-P122G-50UG 500-P122G-100UG 500-P122G-1MG
10776-166EA 560.17 CAD
10776-166 10776-168 10776-170
Anti-IFN-gamma Goat Polyclonal Antibody
Antibodies
Produced from sera of goats immunized with highly pure Recombinant Rat IFN-γ. Anti­Rat IFN-γ­specific antibody was purified by affinity chromatography employing an immobilized Rat IFN-γ matrix.

Applications tested can vary by lot number. See Product Data Sheet for more information.

Type: Primary
Antigen: IFN-gamma
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: Goat IgG
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR