Anti-Resistin Goat Polyclonal Antibody

Supplier: Peprotech
500-P182G-50UG 500-P182G-100UG 500-P182G-1MG
10776-694EA 536.88 CAD
10776-694 10776-696 10776-698
Anti-Resistin Goat Polyclonal Antibody
Antibodies
Produced from sera of goats immunized with highly pure Recombinant Murine Resistin. Anti­Murine Resistin­specific antibody was purified by affinity chromatography employing an immobilized Murine Resistin matrix.

Applications tested can vary by lot number. See Product Data Sheet for more information.

Type: Primary
Antigen: Resistin
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: Goat IgG
Reactivity: Murine
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR