Anti-M-CSF Goat Polyclonal Antibody

Supplier: Peprotech
500-P62G-50UG 500-P62G-100UG 500-P62G-1MG
10781-516EA 565.59 CAD
10781-516 10781-518 10781-520
Anti-M-CSF Goat Polyclonal Antibody
Antibodies
Produced from sera of goats immunized with highly pure Recombinant Murine M-CSF. Anti­Murine M-CSF­specific antibody was purified by affinity chromatography employing an immobilized Murine M-CSF matrix.

Applications tested can vary by lot number. See Product Data Sheet for more information.

Type: Primary
Antigen: M-CSF
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: Goat IgG
Reactivity: Murine
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR