Anti-IL-18BP Goat Polyclonal Antibody

Supplier: Peprotech
500-P153G-50UG 500-P153G-100UG 500-P153G-1MG
10776-414EA 565.59 CAD
10776-414 10776-416 10776-418
Anti-IL-18BP Goat Polyclonal Antibody
Antibodies
Produced from sera of goats immunized with highly pure Recombinant Murine IL-18BP. Anti­Murine IL-18BP­specific antibody was purified by affinity chromatography employing an immobilized Murine IL-18BP matrix.

Applications tested can vary by lot number. See Product Data Sheet for more information.

Type: Primary
Antigen: IL-18BP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: Goat IgG
Reactivity: Murine
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR