Anti-IL-12 Goat Polyclonal Antibody

Supplier: Peprotech
500-P155G-50UG 500-P155G-100UG 500-P155G-1MG
10776-438EA 595.93 CAD
10776-438 10776-440 10776-442
Anti-IL-12 Goat Polyclonal Antibody
Antibodies
Produced from sera of goats immunized with highly pure Recombinant Murine IL-12. Anti­Murine IL-12­specific antibody was purified by affinity chromatography employing an immobilized Murine IL-12 matrix.

Applications tested can vary by lot number. See Product Data Sheet for more information.

Type: Primary
Antigen: IL-12
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: Goat IgG
Reactivity: Murine
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR