Anti-TWEAK Goat Polyclonal Antibody

Supplier: Peprotech
500-P137G-50UG 500-P137G-100UG 500-P137G-1MG
10771-400EA 536.88 CAD
10771-400 10771-402 10771-404
Anti-TWEAK Goat Polyclonal Antibody
Antibodies
Produced from sera of goats immunized with highly pure Recombinant Human TWEAK. Anti­Human TWEAK­specific antibody was purified by affinity chromatography employing an immobilized Human TWEAK matrix.

Applications tested can vary by lot number. See Product Data Sheet for more information.

Type: Primary
Antigen: TWEAK
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: Goat IgG
Reactivity: Human
Order Now

SPECIFICATIONS

Antigen symbol TWEAK
Antigen name TWEAK
Environmentally Preferable
Antibody type Primary
Clonality Polyclonal
Conjugation Unconjugated
Reactivity Human
Host Goat
Isotype IgG
Western blot Yes
ELISA Yes
Immunogen Recombinant Human TWEAK
Form Lyophilized
Molecular weight 17.0 kDa
Purification Recombinant Human TWEAK
Storage buffer See Product Data Sheet for more information.
Storage temperature Store at –20 °C prior to reconstitution.
Shipping temperature Ambient

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR