Anti-SCF Goat Polyclonal Antibody

Supplier: Peprotech
500-P48G-50UG 500-P48G-100UG 500-P48G-1MG
10775-386EA 560.17 CAD
10775-386 10775-388 10775-390
Anti-SCF Goat Polyclonal Antibody
Antibodies
Produced from sera of goats immunized with highly pure Recombinant Human SCF. Anti­Human SCF­specific antibody was purified by affinity chromatography employing an immobilized Human SCF matrix.

Applications tested can vary by lot number. See Product Data Sheet for more information.

Type: Primary
Antigen: SCF
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: Goat IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR