Anti-IL-20 Goat Polyclonal Antibody

Supplier: Peprotech
500-P190G-50UG 500-P190G-100UG 500-P190G-1MG
10776-758EA 536.88 CAD
10776-758 10776-760 10776-762
Anti-IL-20 Goat Polyclonal Antibody
Antibodies
Produced from sera of goats immunized with highly pure Recombinant Human IL-20. Anti­Human IL-20­specific antibody was purified by affinity chromatography employing an immobilized Human IL-20 matrix.

Applications tested can vary by lot number. See Product Data Sheet for more information.

Type: Primary
Antigen: IL-20
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: Goat IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR